5-Methylpyrazine-2-carboxylic acid-4-oxide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Acipimox, 5-Carboxy-2-methylpyrazine 1-oxide, 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide
A635376-100MG
A635376-50MG
A635376-10G
A635376-5G
76980-702EA
7.43
USD
76980-702
76980-704
77613-970
77673-728
5-Methylpyrazine-2-carboxylic acid-4-oxide 97%
5-Methylpyrazine-2-carboxylic acid-4-oxide
Precaución: for Research Use Only
Formula:
C₆H₆N₂O₃ MW: 154.13 g/mol |
MDL Number:
MFCD00865757 CAS Number: 51037-30-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...