Methyl-5-fluoro-2-iodobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-fluoro-2-iodobenzoate

A271069-10G A271069-1G A271069-250MG A271069-25G A271069-5G
76977-178EA 255.68 USD
76977-178 76977-180 76977-182 76977-184 76977-186
Methyl-5-fluoro-2-iodobenzoate 97%
Methyl 5-fluoro-2-iodobenzoate

Precaución: for Research Use Only

Formula: C₈H₆FIO₂
MW: 280.0349 g/mol
MDL Number: MFCD12965043
CAS Number: 1202897-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR