2-Fluoro-4-(methylsulfonyl)benzeneboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-(methylsulphonyl)phenylboronic acid

A193854-100MG A193854-1G A193854-250MG A193854-5G
76974-950EA 20 USD
76974-950 76974-952 76974-954 76974-956
2-Fluoro-4-(methylsulfonyl)benzeneboronic acid 95%
2-Fluoro-4-(methylsulfonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₄S
MDL Number: MFCD09027248
CAS Number: 957060-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR