(5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-1-((2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Azilsartan medoxomil
A325842-100MG
A325842-10MG
A325842-1MG
A325842-50MG
A325842-5MG
76978-154EA
19.1
USD
76978-154
76978-156
76978-158
76978-160
76978-162
(5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-1-((2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate 98%
(5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-1-((2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate
Precaución: for Research Use Only
Formula:
C₃₀H₂₄N₄O₈ MW: 568.53 g/mol |
MDL Number:
MFCD19443688 CAS Number: 863031-21-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...