2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl 97%

Supplier: Ambeed
Warning

Synonyms: XPhos, 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl

A167774-100G A167774-10G A167774-1G A167774-250MG A167774-25G A167774-500G A167774-5G
76973-850EA 120 USD
76973-850 76973-852 76973-854 76973-856 76973-858 76973-860 76973-862
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl 97%
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₃₃H₄₉P
MW: 476.72 g/mol
MDL Number: MFCD04117682
CAS Number: 564483-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR