4-Amino-N-((2R,3S)-3-amino-2-hydroxy-4-phenylbutyl)-N-isobutylbenzenesulfonamide 95%

Supplier: Ambeed
Warning

A629875-1G A629875-25G A629875-5G
76980-674EA 16.98 USD
76980-674 76980-676 76980-678
4-Amino-N-((2R,3S)-3-amino-2-hydroxy-4-phenylbutyl)-N-isobutylbenzenesulfonamide 95%
4-Amino-N-((2R,3S)-3-amino-2-hydroxy-4-phenylbutyl)-N-isobutylbenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₀H₂₉N₃O₃S
MW: 391.52 g/mol
MDL Number: MFCD16251177
CAS Number: 169280-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR