4-Fluoro-5-iodo-1H-indazole 97%

Supplier: Ambeed
Warning

A169612-100MG A169612-1G A169612-250MG A169612-5G
76973-928EA 9 USD
76973-928 76973-930 76973-932 77881-307
4-Fluoro-5-iodo-1H-indazole 97%
4-Fluoro-5-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄FIN₂
MDL Number: MFCD11007929
CAS Number: 1082041-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR