5-Bromo-4-fluoro-3-iodo-1H-indazole 98%
Supplier: Ambeed
|
Warning
|
A311098-100MG
A311098-250MG
76977-908EA
6
USD
76977-908
76977-910
5-Bromo-4-fluoro-3-iodo-1H-indazole 98%
5-Bromo-4-fluoro-3-iodo-1H-indazole
Precaución: for Research Use Only
Formula:
C₇H₃BrFIN₂ |
MDL Number:
MFCD11007931 CAS Number: 1082041-88-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...