Afatinib 99%

Supplier: Ambeed
Warning

A149039-25MG A149039-1G A149039-100MG A149039-250MG A149039-5G A149039-25G A149039-50MG
76973-026EA 5 USD
76973-026 77549-848 77611-374 77671-094 77671-096 77881-163 77881-164
Afatinib 99%
Afatinib

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClFN₅O₃
MDL Number: MFCD12407405
CAS Number: 850140-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR