1,4-Bis(2-benzoxazolyl)naphthalene 96%

Supplier: Ambeed
Warning

Synonyms: 1,4-Bis(benzo[d]oxazol-2-yl)naphthalene

A198669-100G A198669-10G A198669-25G A198669-500G A198669-5G
76975-152EA 36.08 USD
76975-152 76975-154 76975-156 76975-158 76975-160
1,4-Bis(2-benzoxazolyl)naphthalene 96%
1,4-Bis(2-benzoxazolyl)naphthalene

Precaución: for Research Use Only

Formula: C₂₄H₁₄N₂O₂
MW: 362.39 g/mol
MDL Number: MFCD00357168
CAS Number: 5089-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR