Orotic acid monohydrate 95%

Supplier: Ambeed
Warning

Synonyms: Orotic acid hydrate, 2,6-Dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid hydrate

A151006-100G A151006-25G A151006-10G A151006-500G
76973-106EA 12 USD
76973-106 77317-912 77611-408 77611-410
Orotic acid monohydrate 95%
Orotic acid monohydrate

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄·1H₂O
MW: 156.10 g/mol (anhy)
MDL Number: MFCD00149398
CAS Number: 50887-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR