2,4-Dichlorobenzoic acid 98%

Supplier: Ambeed
Warning

A163396-1000G A163396-100G A163396-500G
76973-668EA 58.35 USD
76973-668 76973-670 76973-672
2,4-Dichlorobenzoic acid 98%
2,4-Dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
MDL Number: MFCD00002414
CAS Number: 50-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR