Fmoc-Lys(ME,Boc)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Lys(Boc)(Me)-OH, Fmoc-Lys(Me,Boc)-OH

A231420-100MG A231420-5G A231420-1G A231420-250MG
76976-168EA 35 USD
76976-168 77550-500 77568-240 77612-258
Fmoc-Lys(ME,Boc)-OH 95%
Fmoc-Lys(ME,Boc)-OH

Precaución: For research use only.

Formula: C₂₇H₃₄N₂O₆
MW: 482.577 g/mol
MDL Number: MFCD01861332
CAS Number: 951695-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR