2-Bromothieno[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A362986-100MG A362986-1G A362986-250MG
76978-616EA 192.03 USD
76978-616 76978-618 77672-824
2-Bromothieno[3,2-c]pyridine 95%
2-Bromothieno[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrNS
MW: 214.08 g/mol
MDL Number: MFCD17677030
CAS Number: 94226-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR