TG101209 98+%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butyl)-3-((5-methyl-2-((4-(4-methylpiperazin-1-yl)phenyl)amino)pyrimidin-4-yl)amino)benzenesulphonamide

A469558-10MG A469558-1MG A469558-25MG A469558-5MG A469558-50MG
76979-636EA 41.38 USD
76979-636 76979-638 76979-640 76979-642 77673-204
TG101209 98+%
TG101209

Precaución: for Research Use Only

Formula: C₂₆H₃₅N₇O₂S
MW: 509.7 g/mol
MDL Number: MFCD15528085
CAS Number: 936091-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR