7-Methylquinazolin-4(3H)-one 95%

Supplier: Ambeed
Warning

A215736-100MG A215736-5G A215736-250MG
76975-682EA 20.16 USD
76975-682 76975-684 77671-932
7-Methylquinazolin-4(3H)-one 95%
7-Methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.17 g/mol
MDL Number: MFCD00089490
CAS Number: 75844-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR