4-n-Pentylphenyl-4-methylbenzoate 98%

Supplier: Ambeed
Warning

A229576-100G A229576-25G A229576-5G
76976-102EA 58 USD
76976-102 76976-104 76976-106
4-n-Pentylphenyl-4-methylbenzoate 98%
4-n-Pentylphenyl-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₉H₂₂O₂
MW: 282.38 g/mol
MDL Number: MFCD01076322
CAS Number: 50649-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR