5-((5-Fluoro-2-oxoindolin-3-ylidene)methyl)-N-(2-hydroxy-3-morpholinopropyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide 98+%

Supplier: Ambeed
Warning

Synonyms: SU14813

A748613-10MG A748613-1MG A748613-50MG A748613-5MG
76981-348EA 73.21 USD
76981-348 76981-350 76981-352 76981-354
5-((5-Fluoro-2-oxoindolin-3-ylidene)methyl)-N-(2-hydroxy-3-morpholinopropyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide 98+%
5-((5-Fluoro-2-oxoindolin-3-ylidene)methyl)-N-(2-hydroxy-3-morpholinopropyl)-2,4-dimethyl-1H-pyrrole-3-carboxamide

Precaución: for Research Use Only

Formula: C₂₃H₂₇FN₄O₄
MW: 442.48 g/mol
MDL Number: MFCD16495822
CAS Number: 627908-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR