6-Chloro-9-isopropyl-9H-purine 95%

Supplier: Ambeed
Danger

A500794-100MG A500794-250MG
76979-862EA 61.54 USD
76979-862 76979-864
6-Chloro-9-isopropyl-9H-purine 95%
6-Chloro-9-isopropyl-9H-purine

Precaución: for Research Use Only

Formula: C₈H₉ClN₄
MW: 196.63 g/mol
MDL Number: MFCD07440137
CAS Number: 500539-08-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR