tert-Butyl ((1S,3R)-3-hydroxycyclopentyl)carbamate 98%

Supplier: Ambeed
Warning

A119998-100MG A119998-1G A119998-250MG A119998-25G
76971-444EA 15 USD
76971-444 77670-700 77670-702 77880-970
tert-Butyl ((1S,3R)-3-hydroxycyclopentyl)carbamate 98%
tert-Butyl ((1S,3R)-3-hydroxycyclopentyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD11878011
CAS Number: 167465-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR