8-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 95%
Supplier: Ambeed
|
Warning
|
A331165-100MG
A331165-1G
A331165-250MG
A331165-5G
76978-248EA
22.28
USD
76978-248
76978-250
76978-252
76978-254
8-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid 95%
8-Bromoimidazo[1,2-a]pyridine-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₈H₅BrN₂O₂ MW: 241.04 g/mol |
MDL Number:
MFCD12195903 CAS Number: 1026201-45-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...