3,4-Dichloro-6-fluoroaniline 98%

Supplier: Ambeed
Warning

A177074-1G A177074-250MG A177074-5G
76974-280EA 55 USD
76974-280 76974-282 76974-284
3,4-Dichloro-6-fluoroaniline 98%
3,4-Dichloro-6-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂FN
MDL Number: MFCD19707630
CAS Number: 2729-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR