N4,N4-Dimethylpyrimidine-4,6-diamine 95%

Supplier: Ambeed
Warning

A946175-100MG A946175-1G
76982-002EA 99.73 USD
76982-002 76982-004
N4,N4-Dimethylpyrimidine-4,6-diamine 95%
N4,N4-Dimethylpyrimidine-4,6-diamine

Precaución: for Research Use Only

Formula: C₆H₁₀N₄
MW: 138.17 g/mol
MDL Number: MFCD18819667
CAS Number: 36314-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR