2-Hydroxy-5-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A730916-250MG A730916-1G
76981-278EA 32 USD
76981-278 77573-348
2-Hydroxy-5-nitrobenzonitrile 98%
2-Hydroxy-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄N₂O₃
MDL Number: MFCD00234258
CAS Number: 39835-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR