Methyl-3-bromo-4,5-dichlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-bromo-4,5-dichlorobenzoate

A150154-1G A150154-250MG
76973-062EA 53.05 USD
76973-062 76973-064
Methyl-3-bromo-4,5-dichlorobenzoate 98%
Methyl 3-bromo-4,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O₂
MW: 283.9300 g/mol
MDL Number: MFCD11846093
CAS Number: 1160574-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR