Methyl 3-amino-4,5-dichlorobenzoate 95%

Supplier: Ambeed
Warning

A141813-1G A141813-250MG
76972-624EA 122.01 USD
76972-624 76972-626
Methyl 3-amino-4,5-dichlorobenzoate 95%
Methyl 3-amino-4,5-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₇Cl₂NO₂
MW: 220.05 g/mol
MDL Number: MFCD14694721
CAS Number: 1250663-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR