Methyl pyridazine-3-carboxylate 96%

Supplier: Ambeed
Warning

A123678-1G A123678-250MG A123678-5G
76971-696EA 37 USD
76971-696 76971-698 76971-700
Methyl pyridazine-3-carboxylate 96%
Methyl pyridazine-3-carboxylate

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MDL Number: MFCD10698044
CAS Number: 34253-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR