2,4-Dimethylquinazoline 95%

Supplier: Ambeed
Warning

A444126-100MG A444126-1G A444126-250MG
76979-404EA 113.52 USD
76979-404 76979-406 76979-408
2,4-Dimethylquinazoline 95%
2,4-Dimethylquinazoline

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MW: 158.2000 g/mol
MDL Number: MFCD18449069
CAS Number: 703-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR