4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]decan-8-yl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A718116-100MG A718116-1G A718116-250MG A718116-5G
76981-218EA 33.95 USD
76981-218 76981-220 76981-222 76981-224
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]decan-8-yl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(1,4-dioxaspiro[4.5]decan-8-yl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₄H₂₅BO₄
MW: 268.15 g/mol
MDL Number: MFCD14581935
CAS Number: 1106871-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR