5,6-Dihydrouracil 98%

Supplier: Ambeed
Warning

A502863-100G A502863-100MG A502863-1G A502863-250MG A502863-25G A502863-5G
76979-890EA 335 USD
76979-890 76979-892 76979-894 76979-896 76979-898 76979-900
5,6-Dihydrouracil 98%

Precaución: for Research Use Only

Formula: C₄H₆N₂O₂
MDL Number: MFCD00006029
CAS Number: 504-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR