Dipyrazino[2,3-f:2',3'-h]quinoxaline-2,3,6,7,10,11-hexacarbonitrile 95%

Supplier: Ambeed
Warning

A422951-100MG A422951-1G A422951-250MG
76979-262EA 14 USD
76979-262 76979-264 76979-266
Dipyrazino[2,3-f:2',3'-h]quinoxaline-2,3,6,7,10,11-hexacarbonitrile 95%
Dipyrazino[2,3-f:2',3'-h]quinoxaline-2,3,6,7,10,11-hexacarbonitrile

Precaución: for Research Use Only

Formula: C₁₈N₁₂
MDL Number: MFCD19689401
CAS Number: 105598-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR