2-(Trifluoromethyl)pyridine-4-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[2-(trifluoromethyl)pyridin-4-yl]-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2-(trifluoromethyl)pyridine

A234500-100MG A234500-1G A234500-250MG A234500-5G A234500-10G A234500-25G
76976-282EA 9 USD
76976-282 76976-284 76976-286 76976-288 77881-607 77881-608
2-(Trifluoromethyl)pyridine-4-boronic acid, pinacol ester 97%
2-(Trifluoromethyl)pyridine-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BF₃NO₂
MW: 273.059 g/mol
MDL Number: MFCD09607678
CAS Number: 1036990-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR