Ethyl 2-(4-bromo-2-nitrophenyl)acetate 98%

Supplier: Ambeed
Warning

A238474-1G A238474-5G
76976-384EA 21 USD
76976-384 77881-624
Ethyl 2-(4-bromo-2-nitrophenyl)acetate 98%
Ethyl 2-(4-bromo-2-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO₄
MDL Number: MFCD11977365
CAS Number: 199328-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR