5-Methylisoquinolin-1(2H)-one 97%

Supplier: Ambeed
Warning

A704276-1G A704276-250MG A704276-5G
76981-122EA 46 USD
76981-122 76981-124 76981-126
5-Methylisoquinolin-1(2H)-one 97%
5-Methylisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD07357397
CAS Number: 24188-72-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR