5,7-Dichloro-3-isopropylpyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A197616-1G A197616-5G A197616-250MG
76975-112EA 352.22 USD
76975-112 76975-114 77611-896
5,7-Dichloro-3-isopropylpyrazolo[1,5-a]pyrimidine 97%
5,7-Dichloro-3-isopropylpyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₉H₉Cl₂N₃
MW: 230.09 g/mol
MDL Number: MFCD12923044
CAS Number: 771510-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR