5-Bromoquinoline-8-carboxylic acid 98%

Supplier: Ambeed
Warning

A117397-100MG A117397-250MG A117397-5G A117397-10G
76971-282EA 12 USD
76971-282 76971-284 76971-286 77880-947
5-Bromoquinoline-8-carboxylic acid 98%
5-Bromoquinoline-8-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MDL Number: MFCD19689072
CAS Number: 928839-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR