4-(Piperidin-4-yl)morpholine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Piperidinyl)morpholine hydrochloride

A503272-1G A503272-250MG
76979-904EA 147.47 USD
76979-904 76979-906
4-(Piperidin-4-yl)morpholine hydrochloride 98%
4-(Piperidin-4-yl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₉ClN₂O
MW: 206.71 g/mol
MDL Number: MFCD03274733
CAS Number: 550370-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR