Methylsulfonylacetonitrile 98%

Supplier: Ambeed
Warning

A540660-25G A540660-100G
76980-182EA 60.48 USD
76980-182 77594-664
Methylsulfonylacetonitrile 98%
Methylsulfonylacetonitrile

Precaución: for Research Use Only

Formula: C₃H₅NO₂S
MW: 119.14 g/mol
MDL Number: MFCD00014743
CAS Number: 2274-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR