2,2'-Dimethoxy-1,1'-binaphthyl 97%

Supplier: Ambeed
Danger

Synonyms: 2,2'-Dimethoxy-1,1'-binaphthalene

A972104-1G A972104-25G A972104-5G
76982-038EA 27.59 USD
76982-038 76982-040 76982-042
2,2'-Dimethoxy-1,1'-binaphthyl 97%
2,2'-Dimethoxy-1,1'-binaphthyl

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₂
MW: 314.384 g/mol
MDL Number: MFCD00091146
CAS Number: 2960-93-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR