3-Formyl-6-isopropylchromone 98%
Supplier: Ambeed
|
Warning
|
A623714-25G
A623714-100G
A623714-5G
76980-644EA
108
USD
76980-644
77882-451
77882-452
3-Formyl-6-isopropylchromone 98%
3-Formyl-6-isopropylchromone
Precaución: for Research Use Only
Formula:
C₁₃H₁₂O₃ |
MDL Number:
MFCD00192183 CAS Number: 49619-58-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...