Iminodibenzyl 98%

Supplier: Ambeed
Warning

Synonyms: 10,11-Dihydro-5H-dibenzo[b,f]azepine

A647243-100G A647243-25G A647243-1000G A647243-500G
76980-780EA 19.1 USD
76980-780 76980-782 77673-758 77673-760
Iminodibenzyl 98%
Iminodibenzyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃N
MW: 195.27 g/mol
MDL Number: MFCD00005070
CAS Number: 494-19-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR