3-Chloro-5-fluorophenylacetonitrile 95%

Supplier: Ambeed
Danger

Synonyms: 3-Chloro-5-fluorophenylacetonitrile, 2-(3-Chloro-5-fluorophenyl)acetonitrile, 3-Chloro-5-fluorobenzyl cyanide

A863173-1G A863173-5G
76981-842EA 28.65 USD
76981-842 76981-844
3-Chloro-5-fluorophenylacetonitrile 95%
3-Chloro-5-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MW: 169.59 g/mol
MDL Number: MFCD04038259
CAS Number: 493038-93-0
UN: 3276
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR