4-Quinazolone 98%

Supplier: Ambeed
Warning

A203001-25MG A203001-100G A203001-25G A203001-5G
76975-286EA 5.31 USD
76975-286 77611-966 77611-968 77671-800
4-Quinazolone 98%
4-Quinazolone

Precaución: for Research Use Only

Formula: C₈H₆N₂O
MW: 146.15 g/mol
MDL Number: MFCD00511302
CAS Number: 491-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR