Methyl-3-bromo-1H-indazole-6-carboxylate 97%

Supplier: Ambeed
Warning

A652113-1G A652113-5G A652113-10G A652113-25G
76980-824EA 23 USD
76980-824 76980-826 77882-495 77882-496
Methyl-3-bromo-1H-indazole-6-carboxylate 97%
Methyl 3-bromo-1H-indazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MDL Number: MFCD11499017
CAS Number: 192945-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR