Ethyl 2-(6-chloropyridazin-3-yl)acetate 95%

Supplier: Ambeed
Warning

A389443-1G A389443-250MG
76978-928EA 509.24 USD
76978-928 76978-930
Ethyl 2-(6-chloropyridazin-3-yl)acetate 95%
Ethyl 2-(6-chloropyridazin-3-yl)acetate

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₂
MW: 200.62 g/mol
MDL Number: MFCD09881204
CAS Number: 1023817-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR