9-Fluorenone 98%

Supplier: Ambeed
Warning

Synonyms: 9H-Fluoren-9-one

A109314-1000G A109314-100G A109314-25G A109314-500G
76970-644EA 58.35 USD
76970-644 76970-646 76970-648 76970-650
9-Fluorenone 98%
9-Fluorenone

Precaución: for Research Use Only

Formula: C₁₃H₈O
MW: 180.21 g/mol
MDL Number: MFCD00001141
CAS Number: 486-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR