Ethyl-2-bromo-5-oxazolecarboxylate 97%

Supplier: Ambeed
Warning

A112211-100MG A112211-1G A112211-5G A112211-250MG
76970-892EA 29 USD
76970-892 77549-178 77565-966 77610-888
Ethyl-2-bromo-5-oxazolecarboxylate 97%
Ethyl 2-bromo-5-oxazolecarboxylate

Precaución: For research use only.

Formula: C₆H₆BrNO₃
MDL Number: MFCD09743496
CAS Number: 1060816-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR