5-Methoxy-2-sulfanyl-4-pyrimidinol 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2-thioxo-2,3-dihydropyrimidin-4(1H)-one

A222712-5G A222712-25G
76975-878EA 74 USD
76975-878 77881-563
5-Methoxy-2-sulfanyl-4-pyrimidinol 97%
5-Methoxy-2-sulfanyl-4-pyrimidinol

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MW: 158.18 g/mol
MDL Number: MFCD00094470
CAS Number: 6939-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR