(S)-1-(3,5-Difluorophenyl)propan-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A514385-100MG A514385-1G A514385-250MG
76979-944EA 172.93 USD
76979-944 76979-946 76979-948
(S)-1-(3,5-Difluorophenyl)propan-1-amine hydrochloride 95%
(S)-1-(3,5-Difluorophenyl)propan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂ClF₂N
MW: 207.64 g/mol
MDL Number: MFCD11101248
CAS Number: 1212812-49-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR