4-Chloro-2-methoxy-5-nitropyridine 95+%

Supplier: Ambeed
Warning

A326565-100MG A326565-250MG
76978-180EA 59.42 USD
76978-180 76978-182
4-Chloro-2-methoxy-5-nitropyridine 95+%
4-Chloro-2-methoxy-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₃
MW: 188.56 g/mol
MDL Number: MFCD19689147
CAS Number: 955395-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR